2-Bromo-6-nitro-4-(trifluoromethoxy)aniline 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-4-(trifluoromethoxy)-6-nitroaniline
A823880-1G
A823880-25G
A823880-5G
76776-618EA
10
USD
76776-618
76776-620
76776-622
2-Bromo-6-nitro-4-(trifluoromethoxy)aniline 96%
2-Bromo-6-nitro-4-(trifluoromethoxy)aniline
Precaución: for Research Use Only
Formula:
C₇H₄BrF₃N₂O₃ |
MDL Number:
MFCD04039212 CAS Number: 886499-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...