6-Undecanamine 98%

Supplier: Ambeed
Danger

A819361-1G A819361-250MG A819361-25G A819361-5G
76776-610EA 47 USD
76776-610 76776-612 76776-614 77878-670
6-Undecanamine 98%
6-Undecanamine

Precaución: for Research Use Only

Formula: C₁₁H₂₅N
MDL Number: MFCD00047924
CAS Number: 33788-00-0
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR