3-(Pyridin-4-yl)isoxazol-5-amine 98%

Supplier: Ambeed
Warning

A819281-100MG A819281-250MG A819281-5G A819281-1G
76776-604EA 22.28 USD
76776-604 76776-606 76776-608 77599-114
3-(Pyridin-4-yl)isoxazol-5-amine 98%
3-(Pyridin-4-yl)isoxazol-5-amine

Precaución: for Research Use Only

Formula: C₈H₇N₃O
MW: 161.16 g/mol
MDL Number: MFCD07787187
CAS Number: 19790-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR