1-Chloro-3,5-dimethyladamantane 98%

Supplier: Ambeed
Warning

A818218-1G A818218-250MG A818218-25G A818218-5G
76776-590EA 35.01 USD
76776-590 76776-592 76776-594 76776-596
1-Chloro-3,5-dimethyladamantane 98%
1-Chloro-3,5-dimethyladamantane

Precaución: for Research Use Only

Formula: C₁₂H₁₉Cl
MW: 198.73 g/mol
MDL Number: MFCD00074841
CAS Number: 707-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR