2-Amino-5-(trifluoromethyl)-1H-benzimidazole 97%

Supplier: Ambeed
Warning

A814659-1G A814659-250MG A814659-100MG A814659-5G
76776-574EA 112 USD
76776-574 76776-576 77878-660 77878-661
2-Amino-5-(trifluoromethyl)-1H-benzimidazole 97%
2-Amino-5-(trifluoromethyl)-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₈H₆F₃N₃
MDL Number: MFCD06659792
CAS Number: 10057-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR