5-Fluoro-2-methoxycinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(5-Fluoro-2-methoxyphenyl)acrylic acid

A809583-100MG A809583-1G A809583-250MG
76776-562EA 24.41 USD
76776-562 76776-564 76776-566
5-Fluoro-2-methoxycinnamic acid 98%
5-Fluoro-2-methoxycinnamic acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₃
MW: 196.18 g/mol
MDL Number: MFCD03002811
CAS Number: 157518-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR