3-Amino-5-phenyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

A806477-100G A806477-10G A806477-1G A806477-25G A806477-5G
76776-550EA 109 USD
76776-550 76776-552 76776-554 76776-556 76776-558
3-Amino-5-phenyl-1H-pyrazole 97%
3-Amino-5-phenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₉N₃
MDL Number: MFCD00191749
CAS Number: 1572-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR