7-Methyl-8-nitroquinoline 98%

Supplier: Ambeed
Warning

A803106-1G A803106-250MG A803106-5G
76776-536EA 50 USD
76776-536 76776-538 76776-540
7-Methyl-8-nitroquinoline 98%
7-Methyl-8-nitroquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00041234
CAS Number: 7471-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR