4-Phenylsulfanylbutyric acid 97%

Supplier: Ambeed
Warning

A802764-1G A802764-250MG
76776-532EA 66 USD
76776-532 76776-534
4-Phenylsulfanylbutyric acid 97%
4-Phenylsulfanylbutyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂S
MDL Number: MFCD02082831
CAS Number: 17742-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR