6-Bromopicolinamide 98%

Supplier: Ambeed
Warning

A796457-1G A796457-250MG A796457-5G
76776-512EA 201.58 USD
76776-512 76776-514 76776-516 77878-638
6-Bromopicolinamide 98%
6-Bromopicolinamide

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O
MDL Number: MFCD00234154
CAS Number: 25194-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR