Ethyl (4-chloro-3-(trifluoromethyl)phenyl)carbamate 97%
Supplier: Ambeed
|
Warning
|
A791705-250MG
A791705-1G
76776-496EA
24
USD
76776-496
77627-578
Ethyl (4-chloro-3-(trifluoromethyl)phenyl)carbamate 97%
Ethyl (4-chloro-3-(trifluoromethyl)phenyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₀H₉ClF₃NO₂ MW: 267.63 g/mol |
MDL Number:
MFCD00043450 CAS Number: 18585-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...