Cinnamyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-1-phenyl-1-propene

A790627-25G A790627-5G
76776-492EA 6.37 USD
76776-492 76776-494
Cinnamyl chloride 95%
Cinnamyl chloride

Precaución: for Research Use Only

Formula: C₉H₉Cl
MW: 152.62 g/mol
MDL Number: MFCD00000986
CAS Number: 2687-12-9
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR