N-Benzyl-2-chloroacetamide 98%

Supplier: Ambeed
Warning

A784869-10G A784869-1G A784869-5G
76776-468EA 182.48 USD
76776-468 76776-470 76776-472
N-Benzyl-2-chloroacetamide 98%
N-Benzyl-2-chloroacetamide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
MDL Number: MFCD00084929
CAS Number: 2564-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR