5-Bromo-3-methylthiophene-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A784546-100MG A784546-1G A784546-250MG A784546-5G
76776-460EA 27.59 USD
76776-460 76776-462 76776-464 76776-466
5-Bromo-3-methylthiophene-2-carboxylic acid 95%
5-Bromo-3-methylthiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅BrO₂S
MDL Number: MFCD11040352
CAS Number: 38239-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR