6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid 95%

Supplier: Ambeed
Warning

Synonyms: Trolox, 6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid

A783985-1MG A783985-250MG A783985-1G A783985-5G A783985-100G
76776-454EA 6 USD
76776-454 76776-456 77603-010 77603-012 77627-562
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid 95%
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid

Precaución: for Research Use Only

Formula: C₁₄H₁₈O₄
MW: 250.29 g/mol
MDL Number: MFCD00006846
CAS Number: 53188-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR