6-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline 97%

Supplier: Ambeed
Danger

A783716-1G A783716-250MG
76776-450EA 51.99 USD
76776-450 76776-452
6-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline 97%
6-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₃NO
MDL Number: MFCD00153078
CAS Number: 1701-22-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR