3-Fluoro-5-(trifluoromethyl)benzyl bromide 98%

Supplier: Ambeed
Danger

A782578-1G A782578-250MG A782578-25G A782578-5G
76776-438EA 29 USD
76776-438 76776-440 76776-442 76776-444
3-Fluoro-5-(trifluoromethyl)benzyl bromide 98%
3-Fluoro-5-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₅BrF₄
MW: 257.03
Boiling Pt: 43
MDL Number: MFCD00061175
CAS Number: 239087-09-3
UN: 1760
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR