4-Propylcyclohexanol (cis- and trans- mixture) 98%

Supplier: Ambeed
Danger

A781299-25G A781299-5G
76776-436EA 66 USD
76776-436 77627-548
4-Propylcyclohexanol (cis- and trans- mixture) 98%
4-Propylcyclohexanol (cis- and trans- mixture)

Precaución: for Research Use Only

Formula: C₉H₁₈O
MDL Number: MFCD00091618
CAS Number: 52204-65-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR