Dibenzothiophene 98%

Supplier: Ambeed
Danger

A780328-100G A780328-10G A780328-25G A780328-500G A780328-1000G
76776-426EA 53 USD
76776-426 76776-428 76776-430 76776-432 76776-434 77878-630
Dibenzothiophene 98%
Dibenzothiophene

Precaución: for Research Use Only

Formula: C₁₂H₈S
MDL Number: MFCD00004969
CAS Number: 132-65-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR