2-(3-Chloropropyl)isoindoline-1,3-dione 98%

Supplier: Ambeed
Danger

A778952-1G A778952-250MG
76776-422EA 16.98 USD
76776-422 76776-424
2-(3-Chloropropyl)isoindoline-1,3-dione 98%
2-(3-Chloropropyl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClNO₂
MW: 223.65 g/mol
MDL Number: MFCD00158628
CAS Number: 42251-84-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR