Isoscopoletin (6-hydroxy-4-methylcoumarin) 97%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxy-4-methyl-2H-chromen-2-one, 6-Hydroxy-4-methylcoumarin

A778394-1G A778394-25G A778394-5G
76776-410EA 17 USD
76776-410 76776-412 76776-414
Isoscopoletin (6-hydroxy-4-methylcoumarin) 97%
Isoscopoletin (6-hydroxy-4-methylcoumarin)

Precaución: for Research Use Only

Formula: C₁₀H₈O₃
MW: 176.17 g/mol
MDL Number: MFCD00016967
CAS Number: 2373-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR