m-Terphenyl 99.5%

Supplier: Ambeed
Danger

A777923-1G A777923-25G A777923-5G
76776-398EA 7 USD
76776-398 76776-400 76776-402
m-Terphenyl 99.5%
m-Terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₄
MDL Number: MFCD00003059
CAS Number: 92-06-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR