5-Nitro-1-benzothiophene-2-carboxylic acid 96%

Supplier: Ambeed
Warning

A776888-100MG A776888-10G A776888-1G A776888-250MG A776888-5G
76776-386EA 7 USD
76776-386 76776-388 76776-390 76776-392 77878-621
5-Nitro-1-benzothiophene-2-carboxylic acid 96%
5-Nitro-1-benzothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅NO₄S
MDL Number: MFCD01159700
CAS Number: 6345-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR