5-Cyano-1-pentyne 98%

Supplier: Ambeed
Danger

Synonyms: 5-Hexynenitrile

A774207-1G A774207-250MG A774207-25G A774207-5G
76776-372EA 47.75 USD
76776-372 76776-374 76776-376 76776-378
5-Cyano-1-pentyne 98%
5-Cyano-1-pentyne

Precaución: for Research Use Only

Formula: C₆H₇N
MW: 93.13 g/mol
MDL Number: MFCD00001978
CAS Number: 14918-21-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR