Methyl-5-methoxy-3-oxovalerate 97% (by GC)

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate, 3-Keto-5-methoxyvaleric Acid Methyl Ester, Methyl 5-methoxy-3-oxopentanoate

A773163-100MG A773163-1G A773163-250MG A773163-5G
76776-358EA 16.98 USD
76776-358 76776-360 76776-362 76776-364
Methyl-5-methoxy-3-oxovalerate 97% (by GC)
Methyl-5-methoxy-3-oxovalerate

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
MDL Number: MFCD00042840
CAS Number: 62462-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR