Ethyl (R)-2-hydroxy-4-phenylbutyrate 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Hydroxy-4-phenylbutyric Acid Ethyl Ester

A772401-10G A772401-1G A772401-25G A772401-5G
76776-350EA 26.53 USD
76776-350 76776-352 76776-354 76776-356
Ethyl (R)-2-hydroxy-4-phenylbutyrate 98%
Ethyl (R)-2-hydroxy-4-phenylbutyrate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
MDL Number: MFCD00077794
CAS Number: 90315-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR