5-(2-Nitrophenyl)oxazole 97%

Supplier: Ambeed
Warning

A767495-1G A767495-250MG A767495-5G
76776-336EA 119.89 USD
76776-336 76776-338 76776-340
5-(2-Nitrophenyl)oxazole 97%
5-(2-Nitrophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MW: 190.1555 g/mol
MDL Number: MFCD00085146
CAS Number: 89808-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR