5-(Aminomethyl)-2-methylpyrimidin-4-amine dihydrochloride 95%

Supplier: Ambeed
Warning

A767213-1G A767213-250MG A767213-5G
76776-330EA 27 USD
76776-330 76776-332 76776-334
5-(Aminomethyl)-2-methylpyrimidin-4-amine dihydrochloride 95%
5-(Aminomethyl)-2-methylpyrimidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂Cl₂N₄
MW: 211.09 g/mol
MDL Number: MFCD00058194
CAS Number: 874-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR