5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: XGLUC Sodium Salt, 5-Bromo-4-chloro-3-indolyl-b-D-glucuronide sodium salt

A765714-100MG A765714-250MG
76776-318EA 56.23 USD
76776-318 76776-320
5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt 98%
5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide sodium salt

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrClNO₇
MW: 444.59 g/mol
MDL Number: MFCD00135782
CAS Number: 129541-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR