Methyl 5-(hydroxymethyl)picolinate 95%

Supplier: Ambeed
Warning

A763140-100MG A763140-1G A763140-250MG
76776-312EA 40 USD
76776-312 76776-314 76776-316
Methyl 5-(hydroxymethyl)picolinate 95%
Methyl 5-(hydroxymethyl)picolinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD11848099
CAS Number: 39977-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR