5-(Diethylamino)-2-pentanone 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Diethylamino)pentan-2-one

A762728-250MG A762728-5G
76776-308EA 8.49 USD
76776-308 76776-310
5-(Diethylamino)-2-pentanone 98%
5-(Diethylamino)-2-pentanone

Precaución: for Research Use Only

Formula: C₉H₁₉NO
MW: 157.26 g/mol
MDL Number: MFCD00009058
CAS Number: 105-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR