3-Amino-2-(2,4-difluorophenoxy)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,4-Difluorophenoxy)pyridin-3-amine

A762142-1G A762142-250MG A762142-5G
76776-302EA 50.93 USD
76776-302 76776-304 76776-306
3-Amino-2-(2,4-difluorophenoxy)pyridine 98%
3-Amino-2-(2,4-difluorophenoxy)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₈F₂N₂O
MW: 222.2 g/mol
MDL Number: MFCD00067802
CAS Number: 175135-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR