5,6,7,8-Tetrahydronaphthalene-2-sulfonyl chloride 98%

Supplier: Ambeed
Danger

A761582-100MG A761582-1G A761582-250MG A761582-5G
76776-294EA 35.01 USD
76776-294 76776-296 76776-298 76776-300
5,6,7,8-Tetrahydronaphthalene-2-sulfonyl chloride 98%
5,6,7,8-Tetrahydronaphthalene-2-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂S
MDL Number: MFCD06655218
CAS Number: 61551-49-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR