3-Amino-o-cresol 98%

Supplier: Ambeed
Warning

A760712-1G A760712-10G A760712-5G
76776-290EA 24 USD
76776-290 77878-603 77878-604
3-Amino-o-cresol 98%
3-Amino-o-cresol

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD00007787
CAS Number: 53222-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR