5-(Aminomethyl)furan-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A760701-100MG A760701-250MG
76776-286EA 95.49 USD
76776-286 76776-288
5-(Aminomethyl)furan-2-carboxylic acid 95%
5-(Aminomethyl)furan-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MW: 141.12 g/mol
MDL Number: MFCD04358393
CAS Number: 934-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR