5,7-Dibromoindole 95%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dibromo-1H-indole

A760587-100MG A760587-1G A760587-250MG A760587-5G
76776-278EA 22.28 USD
76776-278 76776-280 76776-282 76776-284
5,7-Dibromoindole 95%
5,7-Dibromoindole

Precaución: for Research Use Only

Formula: C₈H₅Br₂N
MW: 274.94 g/mol
MDL Number: MFCD03090642
CAS Number: 36132-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR