5-Chloro-3-methyl-1H-indole 95%

Supplier: Ambeed
Warning

A760328-100MG A760328-1G A760328-250MG A760328-5G
76776-270EA 6 USD
76776-270 76776-272 76776-274 76776-276
5-Chloro-3-methyl-1H-indole 95%
5-Chloro-3-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈ClN
MDL Number: MFCD04972055
CAS Number: 71095-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR