2-(5-Nitro-2-pyridinyl)acetonitrile 95%

Supplier: Ambeed
Warning

A759411-100MG A759411-1G A759411-250MG A759411-5G
76776-264EA 7 USD
76776-264 76776-266 76776-268 77878-599
2-(5-Nitro-2-pyridinyl)acetonitrile 95%
2-(5-Nitro-2-pyridinyl)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD09033814
CAS Number: 123846-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR