5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane 97%

Supplier: Ambeed
Warning

A759330-100G A759330-25G A759330-5G
76776-258EA 96 USD
76776-258 76776-260 76776-262
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane 97%
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane

Precaución: for Research Use Only

Formula: C₂₁H₂₄O₄
MDL Number: MFCD00021235
CAS Number: 77-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR