4-Morpholin-4-ylbutanenitrile 95%

Supplier: Ambeed
Warning

A757174-1G A757174-25G A757174-5G
76776-248EA 23 USD
76776-248 76776-250 76776-252
4-Morpholin-4-ylbutanenitrile 95%
4-Morpholin-4-ylbutanenitrile

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O
MDL Number: MFCD00023393
CAS Number: 5807-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR