4-Amino-3-hydroxy-N,N-dimethylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

A754719-1G A754719-25G A754719-5G
76776-230EA 30.77 USD
76776-230 76776-232 76776-234
4-Amino-3-hydroxy-N,N-dimethylbenzenesulfonamide 98%
4-Amino-3-hydroxy-N,N-dimethylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₃S
MDL Number: MFCD00036092
CAS Number: 41608-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR