5-Nitroisoindoline hydrochloride 96%

Supplier: Ambeed
Warning

A748029-100MG A748029-250MG A748029-5G A748029-1G
76776-200EA 17 USD
76776-200 76776-202 76776-204 77878-592
5-Nitroisoindoline hydrochloride 96%
5-Nitroisoindoline hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MDL Number: MFCD08234716
CAS Number: 400727-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR