Ethyl-2,6-dimethylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2,6-dimethylbenzoate

A747681-1G A747681-5G A747681-25G
76776-194EA 12.74 USD
76776-194 76776-196 77627-494
Ethyl-2,6-dimethylbenzoate 97%
Ethyl 2,6-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00051785
CAS Number: 36596-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR