H-Thr(Bzl)-OH 98%

Supplier: Ambeed
Warning

Synonyms: O-Benzyl-L-threonine

A746649-5G A746649-1G A746649-25G
76776-192EA 101.85 USD
76776-192 77598-876 77627-492
H-Thr(Bzl)-OH 98%
H-Thr(Bzl)-OH

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.245 g/mol
MDL Number: MFCD00037768
CAS Number: 4378-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR