tert-Butyl 4-(piperazin-1-yl)benzoate 95%

Supplier: Ambeed
Warning

A746556-1G A746556-250MG
76776-188EA 83 USD
76776-188 76776-190
tert-Butyl 4-(piperazin-1-yl)benzoate 95%
tert-Butyl 4-(piperazin-1-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD09608130
CAS Number: 187669-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR