4-Fluoro-2-(trifluoromethyl)benzaldehyde 95%

Supplier: Ambeed
Danger

A743548-10G A743548-1G A743548-5G
76776-166EA 17 USD
76776-166 76776-168 76776-170
4-Fluoro-2-(trifluoromethyl)benzaldehyde 95%
4-Fluoro-2-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₄O
MDL Number: MFCD00061313
CAS Number: 90176-80-0
UN: 1989
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR