Guanidinium hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

A741759-1000G A741759-100G A741759-25G A741759-500G
76776-158EA 15 USD
76776-158 76776-160 76776-162 76776-164
Guanidinium hydrochloride 98%
Guanidinium hydrochloride

Precaución: for Research Use Only

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
MDL Number: MFCD00013026
CAS Number: 50-01-1
Merck Index: 13,04578

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR