Dimethyl(2-piperazin-1-ylethyl)amine 95%

Supplier: Ambeed
Danger

Synonyms: 1-[2-(Dimethylamino)ethyl]piperazine

A739513-1G A739513-5G
76776-154EA 11.67 USD
76776-154 76776-156
Dimethyl(2-piperazin-1-ylethyl)amine 95%
Dimethyl(2-piperazin-1-ylethyl)amine

Precaución: for Research Use Only

Formula: C₈H₁₉N₃
MW: 157.26 g/mol
MDL Number: MFCD00167969
CAS Number: 3644-18-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR