5-Methylpyrazine-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: 2-Carbamoyl-5-methylpyrazine

A738965-1G A738965-250MG A738965-25G A738965-5G
76776-146EA 20.16 USD
76776-146 76776-148 76776-150 76776-152
5-Methylpyrazine-2-carboxamide 98%
5-Methylpyrazine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₇N₃O
MW: 137.14 g/mol
MDL Number: MFCD00190581
CAS Number: 5521-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR