6-Chloro-1H-indole-3-carbonitrile 97%

Supplier: Ambeed
Warning

A737290-100MG A737290-1G A737290-250MG
76776-140EA 49.87 USD
76776-140 76776-142 76776-144
6-Chloro-1H-indole-3-carbonitrile 97%
6-Chloro-1H-indole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅ClN₂
MW: 176.60 g/mol
MDL Number: MFCD07776605
CAS Number: 194490-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR